missing translation for 'onlineSavingsMsg'
Läs mer

Tocris Bioscience™ Canertinib dihydrochloride
GREENER_CHOICE

Produktkod. 18718248
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18718248 25 mg
18772469 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18718248 Leverantör Tocris Bioscience™ Leverantörsnummer 5916/25

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Potent EGFR kinase inhibitor

Canertinib dihydrochloride is a potent EGFR kinase inhibitor (IC50 = 7.4 nM). Blocks autophosphorylation of erbB2. Inhibits growth of A431 tumor xenografts in mice. Cell permeable.
TRUSTED_SUSTAINABILITY

Specifica

Kemiskt namn eller material N-[4-[(3-Chloro-4-fluorophenyl)amino]-7-[3-(4-morpholinyl)propoxy]-6-quinazolinyl]-2-propenamide dihydrochloride
CAS 289499-45-2
Analysprocentintervall >98%
Kvantitet 25 mg
Molekylformel C24H25ClFN5O3.2HCl
Formel vikt Observed MW: 558.86
Titolo del prodotto
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.