Learn More
ACROBiosystems Canine TNF-alpha Protein, His Tag
4673.08 SEK - 34313.77 SEK
Specifications
Molecular Weight (g/mol) | 19.2 kDa |
---|---|
Source | Canine TNF-alpha, His Tag (TNA-C52H3) is expressed from human 293 cells (HEK293). It contains AA Val 77 - Leu 233 (Accession # P51742-1). |
Immunogen | Val 77 - Leu 233 |
Storage Requirements | -20°C |
Gene Alias | DIF,TNF-alpha,TNFA,TNFSF2,cachexin,cachectin,TNFα |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17226590
|
ACROBiosystems
TNA-C52H3-1MG |
1 mg |
34313.77 SEK
1mg |
Estimated Shipment: 30-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17216590
|
ACROBiosystems
TNA-C52H3-100UG |
100 μg |
4673.08 SEK
100µg |
Estimated Shipment: 30-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Tumor necrosis factor alpha (TNFα) is a cytokine produced primarily by monocytes and macrophages. It is found in synovial cells and macrophages in the tissues.The primary role of TNFα is in the regulation of immune cells. TNFα is able to induce apoptotic cell death, to induce inflammation, and to inhibit tumorigenesis and viral replication. Dysregulation of TNFα production has been implicated in a variety of human diseases, including major depression, Alzheimer's disease and cancer. Recombinant TNFα is used as an immunostimulant under the INN tasonermin. TNFα can be produced ectopically in the setting of malignancy and parallels parathyroid hormone both in causing secondary hypercalcemia and in the cancers with which excessive production is associated.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 19.2 kDa.Specifications
19.2 kDa | |
Val 77 - Leu 233 | |
DIF,TNF-alpha,TNFA,TNFSF2,cachexin,cachectin,TNFα | |
Unconjugated | |
His Tag | |
Powder |
Canine TNF-alpha, His Tag (TNA-C52H3) is expressed from human 293 cells (HEK293). It contains AA Val 77 - Leu 233 (Accession # P51742-1). | |
-20°C | |
TNF-alpha | |
Canine | |
HEK293 | |
Cytokines&Growth; Factors |