missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cannabinoid receptor 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17978-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes one of two cannabinoid receptors. The cannabinoids, principally delta-9-tetrahydrocannabinol and synthetic analogs, are psychoactive ingredients of marijuana. The cannabinoid receptors are members of the guanine-nucleotide-binding protein coupled receptor family, which inhibit adenylate cyclase activity in a dose-dependent, stereoselective and pertussis toxin-sensitive manner. The two receptors have been found to be involved in the cannabinoid-induced CNS effects experienced by users of marijuana. Multiple transcript variants encoding two different protein isoforms have been described for this gene.Specifications
Cannabinoid receptor 1 | |
Polyclonal | |
Unconjugated | |
CNR1 | |
CANN6, Cannabinoid receptor 1, cannabinoid receptor 1 (brain), CB R, CB1, CB1A, CB1K5, CB1R, CNR, CNR1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1268, 12801, 25248 | |
-20°C | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P20272, P21554, P47746 | |
Cnr1 | |
Cannabinoid receptor 1 Fusion Protein Ag12371 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |