Learn More
Canrenone, MedChemExpress
Product Code. 16464597
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Product Code. Quantity Unit Size
16464597 50 mg 50mg
1 options
Product Code. 16464597 Supplier Medchem Express Supplier No. HYB1438
 Restricted Item
Beräknad leverans: 30-11-2026
MedChemExpress Canrenone (Aldadiene) is an aldosterone antagonist extensively used as a diuretic agent.
Canrenone (Aldadiene) is an aldosterone antagonist extensively used as a diuretic agent.
Specifications
Chemical Name or Material Canrenone
CAS 976-71-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C22H28O3
Quantity 50 mg
Synonym Aldadiene SC9376
Solubility Information DMSO : ≥ 100 mg/mL (293.72 mM)
SMILES C[C@@]12[C@]3(CCC(O3)=O)CC[C@@]1([H])[C@]4([H])C=CC5=CC(CC[C@]5(C)[C@@]4([H])CC2)=O
Molecular Weight (g/mol) 340.46
Formula Weight 340.46
Percent Purity 99.54%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation