Läs mer
Capecitabine, TRC
Produktkod. 30138248
Change view
Kvantitet:
50 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138248 500 mg
18022150 1 g
17999829 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138248 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC175650500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 154361-50-9
Molekylformel C15 H22 F N3 O6
Molekylvikt (g/mol) 359.35
Synonym Capecitabine, Cytidine, 5'-deoxy-5-fluoro-N-[(pentyloxy)carbonyl]- (9CI, ACI), 5'-Deoxy-5-fluoro-N-[(pentyloxy)carbonyl]cytidine (ACI), Carbamic acid, [1-(5-deoxy-β-D-ribofuranosyl)-5-fluoro-1,2-dihydro-2-oxo-4-pyrimidinyl]-, pentyl ester (ZCI), 5'-Deoxy-5-fluoro-N4-(pentyloxycarbonyl)cytidine, Capacetrine, Capecitabine Accord, Capecitabine Medac, Capecitabine SUN, Capecitabine Teva, Capecitibine, Capecytabine, Capiibine, Captabin, Ecansya, N4-Pentyloxycarbonyl-5'-deoxy-5-fluorocytidine, Ro 09-1978, Ro 09-1978/000, Xeloda, Xeltabin
IUPAC-namn pentyl N-[1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-methyloxolan-2-yl]-5-fluoro-2-oxopyrimidin-4-yl]carbamate
LEDER CCCCCOC(=O)NC1=NC(=O)N(C=C1F)[C@@H]2O[C@H](C)[C@@H](O)[C@H]2O
Specifikationer
Kemiskt namn eller material Capecitabine
Kvantitet 500 mg
InChI formel InChI=1S/C15H22FN3O6/c1-3-4-5-6-24-15(23)18-12-9(16)7-19(14(22)17-12)13-11(21)10(20)8(2)25-13/h7-8,10-11,13,20-21H,3-6H2,1-2H3,(H,17,18,22,23)/t8-,10-,11-,13-/m1/s1
Formel vikt 359.1493