missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CAPG (Human) Recombinant Protein
Human CAPG full-length ORF ( AAH00728.1, 1 a.a. - 348 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00000822-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MYTAIPQSGSPFPGSVQDPGLHVWRVEKLKPVPVAQENQGVFFSGDSYLVLHNGPEEVSHLHLWIGQQSSRDEQGACAVLAVHLNTLLGERPVQHREVQGNESDLFMSYFPRGLKYQEGGVESAFHKTSTGAPAAIKKLYQVKGKKNIRATERALNWDSFNTGDCFILDLGQNIFAWCGGKSNILERNKARDLALAIRDSERQGKAQVEIVTDGEEPAEMIQVLGPKPALKEGNPEEDLTADKANAQAAALYKVSDATGQMNLTKVADSSPFALELLISDDCFVLDNGLCGKIYIWKGRKANEKERQAALQVAEGFISRMQYAPNTQVEILPQGRESPIFKQFFKDWK
Specifications
AAH00728.1 | |
capping protein (actin filament), gelsolin-like | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CAPG | |
GST |
822 | |
Wheat germ expression system | |
25 μg | |
AFCP, MCP | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |