missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CAPN3 (Human) Recombinant Protein
Human CAPN3 full-length ORF ( AAH03169, 1 a.a. - 309 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00000825-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MHGNKQHLQKDFFLYNASKARSKTYINMREVSQRFRLPPSEYVIVPSTYEPHQEGEFILRVFSEKRNLSEEVENTISVDRPVKKKKTKPIIFVSDRANSNKELGVDQESEEGKGKTSPDKQKQSPQPQPGSSDQESEEQQQFRNIFKQIAGDDMEICADELKKVLNTVVNKHKDLKTHGFTLESCRSMIALMDTDGSGKLNLQEFHHLWNKIKAWQKIFKHYDTDQSGTINSYEMRNAVNDAGFHLNNQLYDIITMRYADKHMNIDFDSFICCFVRLEGMFRAFHAFDKDGDGIIKLNVLEWLQLTMYA
Specifications
AAH03169 | |
calpain 3, (p94) | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CAPN3 | |
GST |
825 | |
Wheat germ expression system | |
25 μg | |
CANP3, CANPL3, LGMD2, LGMD2A, MGC10767, MGC11121, MGC14344, MGC4403, nCL-1, p94 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |