Läs mer
Capreomycin Sulfate, TRC
Produktkod. 30345240
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30345240 1 g
1 options
Produktkod. 30345240 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC1756751G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Capreomycin Sulfate
CAS 1405-37-4
Molekylformel C24H44N14O12S
Kvantitet 1 g
Synonym Capastat sulfate, Capreomycin Sulfate, Caprocin
InChI formel InChI=1S/C25H44N14O8.H2O4S/c26-4-1-2-11(27)6-17(41)32-8-14-20(43)35-15(9-34-25(30)47)21(44)39-18(13-3-5-31-24(29)38-13)23(46)33-7-12(28)19(42)37-16(10-40)22(45)36-14;1-5(2,3)4/h5,9,11-14,16,18,24,38,40H,1-4,6-8,10,26-29H2,(H,32,41)(H,33,46)(H,35,43)(H,36,45)(H,37,42)(H,39,44)(H3,30,34,47);(H2,1,2,3,4)/b15-9-;/t11-,12?,13-,14-,16-,18?,24?;/m1./s1 andInChI=1S/C25H44N14O7/c1-11-19(41)36-15(9-32-17(40)7-12(27)3-2-5-26)21(43)37-16(10-34-25(30)46)22(44)39-18(14-4-6-31-24(29)38-14)23(45)33-8-13(28)20(42)35-11/h6,10-15,18,24,38H,2-5,7-9,26-29H2,1H3,(H,32,40)(H,33,45)(H,35,42)(H,36,41)(H,37,43)(H,39,44)(H3,30,34,46)/b16-10-/t11-,12-,13?,14-,15-,18?,24?/m1/s1
LEDER C1CN=C(NC1C2C(=O)NC[C@@H](C(=O)NC(C(=O)N[C@H](C(=O)N/C(=C\NC(=O)N)/C(=O)N2)CNC(=O)[C@H](CCCN)N)CO)N)N.OS(=O)(=O)O
IUPAC-namn (3R)-3,6-diamino-N-(((2R,5R,Z)-15-amino-11-((4R)-2-amino-2,3,4,5-tetrahydropyrimidin-4-yl)-2-methyl-3,6,9,12,16-pentaoxo-8-(ureidomethylene)-1,4,7,10,13-pentaazacyclohexadecan-5-yl)methyl)hexanamide sulfuric acid and (3R)-3,6-diamino-N-(((2R,5R,Z)-15-amino-11-((4R)-2-amino-2,3,4,5-tetrahydropyrimidin-4-yl)-2-(hydroxymethyl)-3,6,9,12,16-pentaoxo-8-(ureidomethylene)-1,4,7,10,13-pentaazacyclohexadecan-5-yl)methyl)hexanamide Sulfuric acid salt
Molekylvikt (g/mol) 752.8