missing translation for 'onlineSavingsMsg'
Learn More

Caprylic acid, >99.5%, MP Biomedicals™

Catalog Number p-4403710
Click to view available options
Quantity:
25 mL
100 mL
500 mL
1 L
This item is not returnable. View return policy
124-07-2
C8H16O2
144.214
MFCD00004429
WWZKQHOCKIZLMA-UHFFFAOYSA-N
caprylic acid, n-octanoic acid, octylic acid, n-caprylic acid, octoic acid, n-octylic acid, n-octoic acid, neo-fat 8, 1-heptanecarboxylic acid, enantic acid
379
CHEBI:28837
octanoic acid
CCCCCCCC(=O)O
This item is not returnable. View return policy
124-07-2
C8H16O2
144.214
MFCD00004429
WWZKQHOCKIZLMA-UHFFFAOYSA-N
caprylic acid, n-octanoic acid, octylic acid, n-caprylic acid, octoic acid, n-octylic acid, n-octoic acid, neo-fat 8, 1-heptanecarboxylic acid, enantic acid
379
CHEBI:28837
octanoic acid
CCCCCCCC(=O)O

Caprylic acid is the common name for the eight-carbon saturated fatty acid known by the systematic name octanoic acid. It is found naturally in the milk of various mammals, and it is a minor constituent of coconut oil and palm kernel oil

Color Colorless
Density 0.91g/cm3 at 20°C
Assay Percent Range >99.5%
Quantity 25 mL
Formula Weight 144.2
Physical Form Liquid
Chemical Name or Material Caprylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'