Läs mer
Captafol, SPEX CertiPrep™
Produktkod. 16350448 Handla allt SPEX Certiprep Produkter
Change view
Kvantitet:
1 mL
Produktkod. Kvantitet
16350448 1 mL
1 options
Produktkod. 16350448 Leverantör SPEX Certiprep Leverantörsnummer S715
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Innehåller
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
AnalyteCAS No.Concentration
Captafol2425-06-11000μg/mL
Certifikat
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Uppfyllande
GHS/OSHA Complaint
Specifikationer
Kemiskt namn eller material Captafol
Förpackning 2mL amber prescored glass ampule
CAS 2425-06-1, 67-64-1
CAS Min % 0.1, 99.9
Anteckningar om koncentration eller sammansättning Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
CAS Max % 0.1, 99.9
Metoder For US EPA Method 8081
Molekylformel C10H9Cl4NO2S
För användning med (utrustning) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
InChI-nyckel JHRWWRDRBPCWTF-UHFFFAOYNA-N
LEDER C1C=CCC2C1C(=O)N(C2=O)SC(C(Cl)Cl)(Cl)Cl
IUPAC-namn 2-(1,1,2,2-tetrachloroethylsulfanyl)-3a,4,7,7a-tetrahydroisoindole-1,3-dione
Lösningsmedel eller matris Acetone
Molekylvikt (g/mol) 349.05
PubChem CID 17038
Kvantitet 1 mL
ChEBI CHEBI:81938
Visa mer Visa mindre
Product intended for laboratory use only