Läs mer
Captafol, TRC
Produktkod. 30344266
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30344266 10 mg
1 options
Produktkod. 30344266 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC17574310MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Captafol
CAS 2425-06-1
Molekylformel C10 H9 Cl4 N O2 S
Kvantitet 10 mg
Synonym 4-Cyclohexene-1,2-dicarboximide, N-[(1,1,2,2-tetrachloroethyl)thio]- (7CI,8CI), 3a,4,7,7a-Tetrahydro-2-[(1,1,2,2-tetrachloroethyl)thio]-1H-isoindole-1,3(2H)-dione, Alfloc 7020, Alfloc 7046, Arborseal, CS 5623, Captafol, Difolatan, Difolatan 4F, Difolatan 4F1, Difolatan 80W, Difolatan BOW, Folcid, Foltaf, Haipen 50, Merpafol, N-(1,1,2,2-Tetrachloroethylthio)-4-cyclohexene-1,2-dicarboximide, N-(1,1,2,2-Tetrachloroethylthio)-4-cyclohexene-1,2-dicarboxyimide, N-(1,1,2,2-Tetrachloroethylthio)-δ4-tetrahydrophthalimide, N-(Tetrachloroethylthio)tetrahydrophthalimide, N-1,1,2,2-Tetrachloroethylmercapto-4-cyclohexene-1,2-dicarboximide, Nalco 7046, Ortho 5865, Proxel EF, Sanspor, Santar SM, Terrazol, Tetrachloroethylthiotetrahydrophthalimide
InChI formel InChI=1S/C10H9Cl4NO2S/c11-9(12)10(13,14)18-15-7(16)5-3-1-2-4-6(5)8(15)17/h1-2,5-6,9H,3-4H2
LEDER ClC(Cl)C(Cl)(Cl)SN1C(=O)C2CC=CCC2C1=O
IUPAC-namn 2-(1,1,2,2-tetrachloroethylsulfanyl)-3a,4,7,7a-tetrahydroisoindole-1,3-dione
Molekylvikt (g/mol) 349.06
Formel vikt 346.9108