missing translation for 'onlineSavingsMsg'
Learn More

Erlab™ Captair™ Smart 321 Ductless Fume Hood

Designed with advanced carbon filtration to minimize chemical exposure.

Brand:  Erlab™ A60B/BE

Additional Details : Weight : 80.00000kg

 View more versions of this product

Product Code. 15514530

  • 36669.66 SEK / Each


Call for latest availability.
+46 (0)31 352 32 30

Add to basket

Description

Description

  • Sensors to detect filter breakthrough of solvents, acids or formaldehyde
  • Safety back up filter in case of main filter saturation
  • Continuous monitoring of air face velocity

Erlab Safety Program

  • Application analysis and validation, usage certification, filter change reminders
  • Connect with eGuard™ for real-time status, notifications and alerts
  • Smart-Light feature for lighting up the hood

Flexibility

  • Configurable filtration column
  • No ductwork needed


Environment

  • No dangerous chemicals are released into atmosphere
  • Low energy consumption
Specifications

Specifications

135 cfm
220V 50/60Hz, 65w
For organic vapor, acid
Filters
Ductless filter hood
61.89 cm
Class2, Type 2C, carbon filter, liquid chemical handling
120.5 cm
80.1 cm