missing translation for 'onlineSavingsMsg'
Learn More

Erlab™ Captair™ Smart 321 Ductless Filtering Fume Hood

Designed with advanced carbon filtration to minimize chemical exposure.

Brand:  Erlab™ A60A/AS

Additional Details : Weight : 70.00000kg

 View more versions of this product

Product Code. 15564520

  • 32002.00 SEK / Each


Estimated Shipment
08-01-2021
Add to basket

Description

Description

  • Smart technology helps user to focus on all chemicals handlings
  • Filtration fume hood equipped with exclusive FLEX modular filtration column allowing use of particulate HEPA or ULPA filters combined with high performance activated carbon molecular filters
  • 7 filtration columns configurations for more versatility
  • Real time sensors to detect main filter saturation with solvents, acids or formaldehyde
  • Safety filter in case of main filter saturation
  • Optional mobile kart or fixed bench
  • Connectable for real time monitoring of all safety parameters
  • Free pre-sale analyze to determine the best filter colum and detector configuration
  • Meets AFNOR NFX 15 211/ANSI Z9.5-2012 filtration efficiency standard (class 1 and 2)
Specifications

Specifications

135 cfm
Ductless filter hood
For organic vapors
Filters
220V 50/60Hz, 65w
61.89 cm
Class2, Type 1c, Liquid chemical handling
111.12 cm
80.1 cm
Videos
SDS
Documents