Learn More
Erlab™ Captair Smart Filtered Storage Cabinets
New Smart Technology uses simple light to show that the hood is operating safely so the user can focus their attention on what is most important: their work. When the light is still the hood is operating safely. To alert the user, the light will pulse associated to an acoustic alarm.
Brand:  Erlab™ B03IA02B
Additional Details : Weight : 40.00000kg
 View more versions of this product
Product Code. 15500731
  • 26657.05 SEK / Each
Call for latest availability.
+46 (0)31 352 32 30
Add to basket
Description
Description
• A simple light pattern will indicate if: air flow is compromised, fan failure, filter breakthrough
• The new eGuard App gives you remote control to monitor the hood, change the settings, and delivers safety alerts immediately to your mobile, tablet or PC device.
• Configurable filtering system above the enclosure can be adapted to filter gases, solvents, powders and particulates keeping the user and the lab protected by safely and efficiently recirculating air within the room and releasing purified air back into the room free of any toxic hazardous vapours or odors
• Meets AFNOR NFX 15 211/ANSI Z9.5-2012 filtration efficiency standard (classes 1
and 2)
• New sensors are added for detection of solvents or acids or formaldehyde
• New electronic anemometer monitors face velocity
• ESP Program: Usage certification delivered with each unit after approval of handlings or process by the manufacturer
• No ducting to the building ventilation system required: Eliminates the energy consumption linked to the air renewal (air supply/extraction, filtration, dehumidification, air conditioning)
• No direct emission of pollutants into the atmosphere
• Very low energy consumption
• Ergonomic design
• Completely mobile and easy to relocate within your lab or to a new building
• To complete with filtration column adapted to your applications, please contact your local Fisher Scientific representative
Specifications
Specifications
Captair 822 Smart Under Bench
Large drawer and sliding tray (without filter)
65 x 55.1 x 76cm (25.59 x 21.69 x 29.92 in.)
Filtering chemical storage cabinet
Videos