Learn More
Erlab™ Captair Smart Filtering Fume Hood Benchcap
Erlab™ Captair Smart Filtering Fume Hood Benchcap offers fixed bench support for Captair Smart fume hoods.
Brand:  Erlab™ I0562000301
Additional Details : Weight : 40.00000kg
 View more versions of this product
Product Code. 15565184
  • 13652.80 SEK / Each
Estimated Shipment: 26-06-2024
to see stock.
Description
Description
  • For Captair Smart filtering fume hoods
  • Equipped with 4 cylinders allowing the leveling of the device
Specifications
Specifications
Smart 481 Fume Hood
Without Work Surface
Metal
910 mm
Fixed Work Bench