Learn More
Erlab™ Captair Smart Trespa™ Work Surface
Erlab™ Captair Smart Trespa™ Work Surface is designed to be used with the range of Captair Smart Ductless Filtering Fume Hoods.
Brand:  Erlab™ I0161030001
Additional Details : Weight : 17.00000kg
 View more versions of this product
Product Code. 15575154
  • 7379.00 SEK / Each
Estimated Shipment
18-12-2020
Add to basket
Description
Description
  • Made of phenolic resin with ergonomic arm for a comfortable working position
  • High chemical and mechanical resistance
  • Ideal for precision weighing operations
Specifications
Specifications
510 mm
Spill Tray
Work Surface
Smart 391 Fume Hood
Phenolic Resin
900 mm