Learn More
Erlab™ CaptairStore™ and ChemTrap™ Filters
Replaces filters in CaptairStore™ and ChemTrap™ filtration systems. CaptiarStore™ and ChemTrap™ Filters are available in a variety of models designed and optimized for specific filtration applications.
2180.00 SEK - 3730.00 SEK
Produktcode | Marke | Filter Type | For Use With (Equipment) | For Use With (Application) | Preis | Menge & Verfügbarkeit | |||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Filter Type | For Use With (Equipment) | For Use With (Application) | Preis | Menge & Verfügbarkeit | |||
11851824
|
Erlab™
H11112201 |
BE | Captair™ Store™ 832 and ChemTrap™ H402 | Polyvalent for Acid and Organic Vapors |
3730.00 SEK
Each |
Estimated Shipment: 05-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11832944
|
Erlab™
F2AS |
AS | ChemTrap™ H402 | For Organic Vapors |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Descrizione
Replacement and spare filters for CaptiarStore™ and ChemTrap™ filtration systems.ChemTrap™ units have been designed to vent and filter vapours coming from chemicals stored in fireproof safety cabinets. They allow fireproof safety cabinets to be totally autonomous by avoiding any connection to energy consuming air extraction systems.
CaptairStore™ filtered storage cabinets are an ideal solution for any laboratory where many flasks and bottles containing toxic or noxious chemicals pollute the lab air and take up valuable bench space.
Specifica
Spare Filter |