missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAPZA1 Mouse anti-Human, Mouse, Rat, Clone: 10H1F9, Proteintech
Mouse Monoclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | CAPZA1 |
---|---|
Clone | 10H1F9 |
Concentration | 0.85 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16837605
|
Proteintech
66066-1-IG-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16827605
|
Proteintech
66066-1-IG-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
CAPZA1 is a member of the F-actin capping protein alpha subunit family. This gene encodes the alpha subunit of the barbed-end actin binding protein. The protein regulates growth of the actin filament by capping the barbed end of growing actin filaments.Specifications
CAPZA1 | |
0.85 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CAPPA1, CAPZ, CapZ alpha 1, CAPZA1, CAZ1 | |
CAPZA1 | |
IgG1 | |
Protein G | |
Antibody |
10H1F9 | |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
B2GUZ5, P47753, P52907 | |
12340, 691149, 829 | |
CAPZA1 Fusion Protein Ag18841 | |
Primary | |
-20°C | |
CAPZA1 |