Läs mer
Carbamazepine EP Impurity G, Mikromol™
Produktkod. 30049284
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30049284 100 mg
30050567 25 mg
30461341 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30049284 Leverantör Mikromol Leverantörsnummer MM0076.05
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Carbamazepine EP Impurity G
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 10-Bromo-5H-dibenzo[b,f]azepine-5-carboxamide (10-Bromocarbamazepine)
CAS 59690-97-0
Analyt- eller komponentnamn 10-Bromo-5H-dibenzo[b,f]azepine-5-carboxamide (10-Bromocarbamazepine)
Frakt skick Room Temperature
Molekylformel C15 H11 Br N2 O
Certifikat och efterlevnad ISO 17025
Synonym 10-Bromo-5H-dibenzo[b,f]azepine-5-carboxamide, Carbamazepine Imp. G (EP), 10-Bromocarbamazepine, Carbamazepine Impurity G
InChI formel InChI=1S/C15H11BrN2O/c16-12-9-10-5-1-3-7-13(10)18(15(17)19)14-8-4-2-6-11(12)14/h1-9H,(H2,17,19)
LEDER NC(=O)N1c2ccccc2C=C(Br)c3ccccc13
IUPAC-namn 5-bromobenzo[b][1]benzazepine-11-carboxamide
Molekylvikt (g/mol) 315.16
Kvantitet 100 mg
Formel vikt 314.0055 g/mol