Läs mer
Carbazeran (>90%), TRC
Produktkod. 30345753
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30345753 25 mg
1 options
Produktkod. 30345753 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC17579025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Carbazeran
CAS 70724-25-3
Molekylformel C18 H24 N4 O4
Kvantitet 25 mg
Synonym Carbamic acid, N-ethyl-, 1-(6,7-dimethoxy-1-phthalazinyl)-4-piperidinyl ester, Carbamic acid, ethyl-, 1-(6,7-dimethoxy-1-phthalazinyl)-4-piperidinyl ester (9CI), 1-(6,7-Dimethoxyphthalazin-1-yl)piperidin-4-yl ethylcarbamate, Carbazeran, UK 31557
InChI formel InChI=1S/C18H24N4O4/c1-4-19-18(23)26-13-5-7-22(8-6-13)17-14-10-16(25-3)15(24-2)9-12(14)11-20-21-17/h9-11,13H,4-8H2,1-3H3,(H,19,23)
LEDER CCNC(=O)OC1CCN(CC1)c2nncc3cc(OC)c(OC)cc23
IUPAC-namn [1-(6,7-dimethoxyphthalazin-1-yl)piperidin-4-yl] N-ethylcarbamate
Molekylvikt (g/mol) 360.41
Formel vikt 360.1798