missing translation for 'onlineSavingsMsg'
Learn More

Carbazeran (>90%), TRC

Product Code. 30338675
Change view
Click to view available options
Kvantitet:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30338675 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30338675 Supplier Toronto Research Chemicals Supplier No. TRCC17579010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Carbazeran
CAS 70724-25-3
Molekylformel C18 H24 N4 O4
Kvantitet 10 mg
Synonym Carbamic acid, N-ethyl-, 1-(6,7-dimethoxy-1-phthalazinyl)-4-piperidinyl ester, Carbamic acid, ethyl-, 1-(6,7-dimethoxy-1-phthalazinyl)-4-piperidinyl ester (9CI), 1-(6,7-Dimethoxyphthalazin-1-yl)piperidin-4-yl ethylcarbamate, Carbazeran, UK 31557
InChI formel InChI=1S/C18H24N4O4/c1-4-19-18(23)26-13-5-7-22(8-6-13)17-14-10-16(25-3)15(24-2)9-12(14)11-20-21-17/h9-11,13H,4-8H2,1-3H3,(H,19,23)
LEDER CCNC(=O)OC1CCN(CC1)c2nncc3cc(OC)c(OC)cc23
IUPAC-namn [1-(6,7-dimethoxyphthalazin-1-yl)piperidin-4-yl] N-ethylcarbamate
Molekylvikt (g/mol) 360.41
Formel vikt 360.1798
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.