Läs mer
Carbazochrome, TRC
Produktkod. 30364456
Change view
Kvantitet:
250 g
Produktkod. Kvantitet
30364456 250 g
1 options
Produktkod. 30364456 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC175820250G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Adenochrome semicarbazone
CAS 69-81-8
Molekylformel C10H12N4O3
Kvantitet 250 g
Synonym Adenochrome semicarbazone, Hydrazinecarboxamide, 2-(1,2,3,6-tetrahydro-3-hydroxy-1-methyl-6-oxo-5H-indol-5-ylidene)-, 5,6-Indolinedione, 3-hydroxy-1-methyl-, 5-semicarbazone (7CI,8CI), Adrenochrome semicarbazone (6CI), 2-(1,2,3,6-Tetrahydro-3-hydroxy-1-methyl-6-oxo-5H-indol-5-ylidene)hydrazinecarboxamide, 3-Hydroxy-1-methyl-5,6-indolindione 5-semicarbazone, Adchnon, Adedolon, Adrenochrome monosemicarbazone, Adrenostazin, Adrenoxyl, Adrezon, Adrokson, Adroxon, Carbazochrom, Carbazochrome, Cartabes, Cromadrenal, Cromosil, NSC 73742, Sangostasin, Sangostazin
InChI formel InChI=1S/C10H12N4O3/c1-14-4-9(16)5-2-6(12-13-10(11)17)8(15)3-7(5)14/h2-3,9,15-16H,4H2,1H3,(H2,11,17)
LEDER CN1CC(O)C2=CC(=NNC(=O)N)C(=O)C=C12
IUPAC-namn (3,6-dihydroxy-1-methyl-2,3-dihydroindol-5-yl)iminourea
Molekylvikt (g/mol) 236.23
Formel vikt 236.0909