Läs mer
Carbidopa Ethyl Ester Hydrochloride Salt, TRC
Produktkod. 30354529
Change view
Kvantitet:
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30354529 25 mg -
30367249 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30354529 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC17604525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 96115-88-7
Molekylformel C12H19ClN2O4
Molekylvikt (g/mol) 290.74
Synonym alpha-Hydrazinyl-3,4-dihydroxy-alpha-methylbenzenepropanoic Acid Ethyl Ester Hydrochloride, alpha-Hydrazino-3,4-dihydroxy-alpha-methylhydrocinnamic Acid Ethyl Ester Hydrochloride
IUPAC-namn ethyl 3-(3,4-dihydroxyphenyl)-2-hydrazinyl-2-methylpropanoate
LEDER Cl.CCOC(=O)C(C)(Cc1ccc(O)c(O)c1)NN
Specifikationer
Kemiskt namn eller material Carbidopa Ethyl Ester Hydrochloride Salt
Kvantitet 25 mg
InChI formel InChI=1S/C12H18N2O4/c1-3-18-11(17)12(2,14-13)7-8-4-5-9(15)10(16)6-8/h4-6,14-16H,3,7,13H2,1-2H3
Formel vikt 290.1033