Läs mer
Carbocisteine-13C3, TRC
Produktkod. 30356082
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30356082 10 mg
30362779 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30356082 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC17876210MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1391068-19-1
Molekylformel C213C3H9NO4S
Molekylvikt (g/mol) 182.17
Synonym S-Carboxymethyl-L-cysteine-13C3, (2R)-2-Amino-3-[(carboxymethyl)thio]propionic Acid-13C3, (R)-S-(Carboxymethyl)cysteine-13C3, (L)-2-Amino-3-(carboxymethylthio)propionic Acid-13C3, 3-[(Carboxymethyl)thio]-L-alanine-13C3, AHR 3053-13C3, Bronchokod-13C3, Carbocisteine-13C3, Carbocit-13C3, Carbocysteine-13C3, Chilvax-13C3, DF 1794Y-13C3, L-(Carboxymethyl)cysteine-13C3, LJ 206-13C3, Lisil-13C3, Lisomucil-13C3, Loviscol-13C3, Muciclar-13C3, Mucocis-13C3, Mucodyne-13C3, Mucofan-13C3, Mucolase-13C3, Mucolex-13C3, Mucopront-13C3, Mucosol-13C3, Mucotab-13C3, Mukinyl, -13C3 NSC 14156-13C3, PectDrill-13C3, Pectox-13C3, Pulmoclase-13C3, Reomucil-13C3, Rhinathiol-13C3, Rhinatiol-13C3, Rinatiol-13C3, S-(Carboxymethyl)-(R)-cysteine-13C3, S-Carboxylmethyl-L-cysteine-13C3, Siroxyl-13C3, Thiodril-13C3, Transbronchin-13C3
IUPAC-namn (2R)-2-amino-3-(carboxymethylsulfanyl)propanoic acid
LEDER N[C@H]([13C](O)=O)CS[13C]([HH])[13C](O)=O
Specifikationer
Kemiskt namn eller material Carbocisteine-13C3
Kvantitet 10 mg
InChI formel InChI=1S/C5H9NO4S/c6-3(5(9)10)1-11-2-4(7)8/h3H,1-2,6H2,(H,7,8)(H,9,10)/t3-/m0/s1