Läs mer
Carbocisteine Sulfoxide (Mixture of diastereomers), TRC
Produktkod. 17918624
Change view
Kvantitet:
250 mg
2.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17918624 250 mg
17987854 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17918624 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC178780250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5439-87-2
Molekylformel C5 H9 N O5 S
Molekylvikt (g/mol) 195.19
Synonym (S)-Carboxymethyl-L-cysteine (R/S)-Sulphoxide
IUPAC-namn (2R)-2-amino-3-(carboxymethylsulfinyl)propanoic acid
LEDER N[C@@H](CS(=O)CC(=O)O)C(=O)O
Specifikationer
Kemiskt namn eller material (S)-Carboxymethyl-L-cysteine (RS)-Sulfoxide
Kvantitet 250 mg
InChI formel InChI=1S/C5H9NO5S/c6-3(5(9)10)1-12(11)2-4(7)8/h3H,1-2,6H2,(H,7,8)(H,9,10)/t3-,12?/m0/s1
Formel vikt 195.0201