Läs mer
Carbomer 934 (Technical Grade), TRC
Produktkod. 30366515
Change view
Kvantitet:
5 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30366515 5 g
30353407 50 g
30365542 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30366515 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC1782335G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 9007-16-3
Molekylformel C5H10O2
Molekylvikt (g/mol) 102.13
Synonym Synthalen M, Acrypol 934
IUPAC-namn 2-methylbutanoic acid polymer
LEDER O=C(O)C(C)CC
Specifikationer
Kemiskt namn eller material Carbomer 934 (Technical Grade)
Kvantitet 5 g
InChI formel InChI=1S/C5H10O2/c1-3-4(2)5(6)7/h4H,3H2,1-2H3,(H,6,7)