missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ Carbonic Anhydrase IV Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5149544
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Immunogen sequence: GTQVNEGFQP LVEALSNIPK PEMSTTMAES SLLDLLPKEE KLRHYFRYLG SLTTPTCDEK VVWTVFREPI QLHREQILAF SQKLYYDKEQ TVSMKDNVRP LQQLGQRTVI KSGA Highest antigen sequence identity to the following orthologs: Mouse - 61%, Rat - 64%.
Carbonic anhydrases are a large family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. They participate in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva, and gastric acid. They show extensive diversity in tissue distribution and in their subcellular localization. This gene encodes a glycosylphosphatidyl-inositol-anchored membrane isozyme expressed on the luminal surfaces of pulmonary capillaries and proximal renal tubules. Its exact function is not known, however, it may have a role in inherited renal abnormalities of bicarbonate transport.
Specifikationer
| Carbonic Anhydrase IV | |
| Polyclonal | |
| Unconjugated | |
| CA4 | |
| AW456718; CA IV; CA4; CAIV; CA-IV; Car4; Carbonate dehydratase IV; carbonic anhydrase 4; Carbonic anhydrase IV; carbonic dehydratase IV; RP17 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 762 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P22748 | |
| CA4 | |
| Recombinant protein corresponding to Human CA4. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering