Läs mer
Invitrogen™ Carbonic Anhydrase VB Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120567
Beskrivning
Positive test controls include: A-549, Mouse kidney, Mouse lung, Mouse spleen, Rat lung. The target is usually found in the following locations: Mitochondrion. Immunogen sequence: MVVMNSLRVI LQASPGKLLW RKFQIPRFMP ARPCSLYTCT YKTRNRALHP LWESVDLVPG GDRQSPINIR WRDSVYDPGL KPLTISYDPA TCLHVWNNGY SFLVEFEDST DKSVIKGGPL EHNYRLKQFH FHWGAIDAWG SEHTVDSKCF PAELHLVHWN AVRFENFEDA ALEENGLAVI GVFLKLGKHH KELQKLVDTL PSIKHKDALV EFGSFDPSCL MPTCPDYWTY SGSLTTPPLS ESVTWIIKKQ PVEVDHDQLE QFRTLLFTSE GEKEKRMVDN FRPLQPLMNR TVRSSFRHDY VLNVQAKPKP ATSQATP.
Carbonic anhydrases (CAs) are members of a large family of zinc metalloenzymes responsible for catalyzing the reversible hydration of carbon dioxide.CAs show extensive diversity in their distribution and subcellular localization.They are involved in a variety of biological processes, including calcification, bone resorption, respiration, acid-base balance and the formation of aqueous humor, saliva, gastric juice and cerebrospinal fluid. CA VB, also known as carbonate dehydratase VB, is one of two isoforms of CA V. It localizes to the mitochondria and is involved in metabolic processes. CA VB is predominantly expressed in heart, pancreas, lung, placenta, kidney and skeletal muscle. It exhibits highest homology with family member CA VA (the second isoform of CA V); however, unlike CA VA, it is not expressed in the liver, suggesting that it plays a significantly different physiological role.
Specifikationer
| Carbonic Anhydrase VB | |
| Polyclonal | |
| Unconjugated | |
| CA5B | |
| 7330410H16Rik; Ca5b; Car5b; Carbonate dehydratase VB; carbonic anhydrase 5B; carbonic anhydrase 5b, mitochondrial; Carbonic anhydrase VB; carbonic anhydrase VB, mitochondrial; carbonic dehydratase; CarVb; CAVB; CA-VB; D730005F19Rik | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 11238, 302669, 56078 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q66HG6, Q9QZA0, Q9Y2D0 | |
| CA5B, Car5b | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-317 of human CA5B (NP_009151.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |