Läs mer
Invitrogen™ Carbonic Anhydrase XII Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587698
Descripción
Immunogen sequence: APVNGSKWTY FGPDGENSWS KKYPSCGGLL QSPIDLHSDI LQYDASLTPL EFQGYNLSAN KQFLLTNNGH SVKLNLPSDM HIQGLQSRYS ATQLHLHWGN PNDPHGSEHT VSGQHFAAEL HIVHYNSDLY PDASTASNKS EGLAVLAVLI EMGSFNPSYD KIFSHLQHVK YKGQEAFVPG FNIEELLPER TAEYYRYRGS LTTPPCNPTV LWTVFRNPVQ ISQEQLLALE TALYCTHMDD PSPREMINNF RQVQKFDERL VYTSFSQVQV CTAAGLS; Positive Samples: HT-29; Cellular Location: Membrane, Single-pass type I membrane protein.
Carbonic Anhydrase XII is a protein coding gene.
Especificaciones
| Carbonic Anhydrase XII | |
| Polyclonal | |
| Unconjugated | |
| CA12 | |
| 2310047E01Rik; AI314958; CA12; Car12; Carbonate dehydratase XII; carbonic anhydrase 12; Carbonic anhydrase XII; carbonic anyhydrase 12; carbonic dehydratase; CAXII; CA-XII; HsT18816; membrane-bound carbonic anhydrase 12; T18816; tumor antigen HOM-RCC-3.1.3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 363085, 76459, 771 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1.84 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O43570, Q8CI85 | |
| CA12, Car12 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 25-301 of human CA12 (NP_0012091). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.