Läs mer
Carbonochloridic Acid Octadecyl Ester, TRC
Produktkod. 30367574
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30367574 500 mg
1 options
Produktkod. 30367574 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC176870500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Carbonochloridic Acid Octadecyl Ester
CAS 51637-93-5
Molekylformel C19H37ClO2
Kvantitet 500 mg
Synonym Chloro-formic Acid Octadecyl Ester, Octadecyl Chloroformate, Stearyl Chloroformate, Stearyloxycarbonyl Chloride, n-Octadecyl Chloroformate
InChI formel InChI=1S/C19H37ClO2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-22-19(20)21/h2-18H2,1H3
LEDER CCCCCCCCCCCCCCCCCCOC(=O)Cl
IUPAC-namn octadecyl carbonochloridate
Molekylvikt (g/mol) 332.95