Läs mer
Carbovir Monophosphate, TRC
Produktkod. 30356557
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30356557 1 mg
1 options
Produktkod. 30356557 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC1777451MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Carbovir Monophosphate
CAS 144490-73-3
Molekylformel C11H14N5O5P
Kvantitet 1 mg
Synonym rel-2-Amino-1,9-dihydro-9-[(1R,4S)-4-[(phosphonooxy)methyl]-2-cyclopenten-1-yl]-6H-purin-6-one, cis-(±)-2-Amino-1,9-dihydro-9-[4-[(phosphonooxy)methyl]-2-cyclopenten-1-yl]-6H-purin-6-one, Carbovir 5’-Phosphate, Abacavir - In House Impurity, Abacavir Impurities
InChI formel InChI=1S/C11H14N5O5P/c12-11-14-9-8(10(17)15-11)13-5-16(9)7-2-1-6(3-7)4-21-22(18,19)20/h1-2,5-7H,3-4H2,(H2,18,19,20)(H3,12,14,15,17)
LEDER NC1=NC(=O)c2ncn(C3CC(COP(=O)(O)O)C=C3)c2N1
IUPAC-namn [4-(2-amino-6-oxo-3H-purin-9-yl)cyclopent-2-en-1-yl]methyl dihydrogen phosphate
Molekylvikt (g/mol) 327.23
Formel vikt 327.0733