Läs mer
Carboxy Primaquine, TRC
Produktkod. 30132998
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132998 1 g
30007749 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30132998 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC1812601G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 77229-68-6
Molekylformel C15 H18 N2 O3
Molekylvikt (g/mol) 274.32
Synonym 4-[(6-Methoxy-8-quinolinyl)amino]pentanoic Acid, 8-(3-Carboxy-1-methylpropylamino)-6-methoxyquinoline;
IUPAC-namn 4-[(6-methoxyquinolin-8-yl)amino]pentanoic acid
LEDER COc1cc(NC(C)CCC(=O)O)c2ncccc2c1
Specifikationer
Kemiskt namn eller material Carboxy Primaquine
InChI formel InChI=1S/C15H18N2O3/c1-10(5-6-14(18)19)17-13-9-12(20-2)8-11-4-3-7-16-15(11)13/h3-4,7-10,17H,5-6H2,1-2H3,(H,18,19)
Kvantitet 1 g
Formel vikt 274.1317