missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carboxypeptidase A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15836-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This antibody is specific to CPA1.
Three different forms of human pancreatic procarboxypeptidase A have been isolated. This gene encodes a monomeric pancreatic exopeptidase involved in zymogen inhibition.Specifications
Carboxypeptidase A1 | |
Polyclonal | |
Unconjugated | |
CPA1 | |
Carboxypeptidase A1, CPA, CPA1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
109697, 1357, 24269 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P00731, P15085, Q7TPZ8 | |
Cpa1 | |
Carboxypeptidase A1 Fusion Protein Ag8586 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |