Läs mer
Carboxyphosphamide Benzyl Ester, TRC
Produktkod. 30358470
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30358470 100 mg
30367554 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30358470 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC181355100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 37979-67-2
Molekylformel C14H21Cl2N2O4P
Molekylvikt (g/mol) 383.21
Synonym 3-[[Amino[bis(2-chloroethyl)amino]phosphinyl]oxy]propanoic Acid Phenylmethyl Ester, 3-[[Amino[bis(2-chloroethyl)amino]phosphinyl]oxy]propanoic Acid Benzyl Ester
IUPAC-namn benzyl 3-[amino-[bis(2-chloroethyl)amino]phosphoryl]oxypropanoate
LEDER NP(=O)(OCCC(=O)OCc1ccccc1)N(CCCl)CCCl
Specifikationer
Kemiskt namn eller material Carboxyphosphamide Benzyl Ester
Kvantitet 100 mg
InChI formel InChI=1S/C14H21Cl2N2O4P/c15-7-9-18(10-8-16)23(17,20)22-11-6-14(19)21-12-13-4-2-1-3-5-13/h1-5H,6-12H2,(H2,17,20)
Formel vikt 382.0616