Läs mer
Carfilzomib, MedChemExpress
Produktkod. 16494986
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
200 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16494986 100 mg
16470337 50 mg
16456697 5 mg
16423988 10 mg
16460986 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Produktkod. 16494986 Leverantör Medchem Express Leverantörsnummer HY10455
MedChemExpress Carfilzomib (PR-171) is an irreversible proteasome inhibitor with an IC50 of 5 nM in ANBL-6 and RPMI 8226 cells.
Carfilzomib (PR-171) is an irreversible proteasome inhibitor with an IC50 of 5 nM in ANBL-6 and RPMI 8226 cells.
Specifikationer
Kemiskt namn eller material Carfilzomib
Fysisk form Powder
CAS 868540-17-4
Färg White
Kvantitet 100 mg
Anteckningar om renhetsgrad Research
Molekylformel C40H57N5O7
För användning med (applikation) Cancer-Kinase/protease
Synonym PR-171
Löslighetsinformation DMF : ≥ 100 mg/mL (138.91 mM) ∣DMSO : 50 mg/mL (69.45 mM; Need ultrasonic)
LEDER C[C@@]1(C([C@H](CC(C)C)NC([C@@H](NC([C@H](CC(C)C)NC([C@@H](NC(CN2CCOCC2)=O)CCC3=CC=CC=C3)=O)=O)CC4=CC=CC=C4)=O)=O)OC1
Molekylvikt (g/mol) 719.91
Formel vikt 719.91
Procent renhet 97.0%