Läs mer
Carfilzomib, TRC
Produktkod. 30135814
Change view
Kvantitet:
5 mg
10 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135814 5 mg
30009482 250 mg
18002320 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135814 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC1834605MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 868540-17-4
Synonym L-Phenylalaninamide, (αS)-α-[[2-(4-morpholinyl)acetyl]amino]benzenebutanoyl-L-leucyl-N-[(1S)-3-methyl-1-[[(2R)-2-methyl-2-oxiranyl]carbonyl]butyl]-∣L-Phenylalaninamide, (αS)-α-[(4-morpholinylacetyl)amino]benzenebutanoyl-L-leucyl-N-[(1S)-3-methyl-1-[[(2R)-2-methyloxiranyl]carbonyl]butyl]- (9CI)∣(αS)-α-[[2-(4-Morpholinyl)acetyl]amino]benzenebutanoyl-L-leucyl-N-[(1S)-3-methyl-1-[[(2R)-2-methyl-2-oxiranyl]carbonyl]butyl]-L-phenylalaninamide∣Carfilzomib∣Kyprolis∣PR 171
Specifikationer
Renhet >95% (HPLC)
Formatera Neat
Molekylformel C40H57N5O7
MDL-nummer MFCD11040997
Molekylvikt (g/mol) 719.91, 719.92
InChI-nyckel BLMPQMFVWMYDKT-UHFFFAOYNA-N
LEDER CC(C)CC(NC(=O)C(CCC1=CC=CC=C1)NC(=O)CN1CCOCC1)C(=O)NC(CC1=CC=CC=C1)C(=O)NC(CC(C)C)C(=O)C1(C)CO1
IUPAC-namn 4-methyl-N-(1-{[4-methyl-1-(2-methyloxiran-2-yl)-1-oxopentan-2-yl]carbamoyl}-2-phenylethyl)-2-{2-[2-(morpholin-4-yl)acetamido]-4-phenylbutanamido}pentanamide
Produkttyp API