missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CARHSP1 (Human) Recombinant Protein
Human CARHSP1 full-length ORF ( NP_001035941.1, 1 a.a. - 147 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00023589-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MSSEPPPPPQPPTHQASVGLLDTPRSRERSPSPLRGNVVPSPLPTRRTRTFSATVRASQGPVYKGVCKCFCRSKGHGFITPADGGPDIFLHISDVEGEYVPVEGDEVTYKMCSIPPKNEKLQAVEVVITHLAPGTKHETWSGHVISS
Specifications
NP_001035941.1 | |
calcium regulated heat stable protein 1, 24kDa | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CARHSP1 | |
GST |
23589 | |
Wheat germ expression system | |
25 μg | |
CRHSP-24, CSDC1, MGC111446 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |