Learn More
Carl Zeiss™ Reflector Module
FL, EP and P & C
Brand:  Carl Zeiss™ 424931-0000-000
Additional Details : Weight : 0.80000kg
Product Code. 11822402
  • 2172.15 SEK / Each
Estimated Shipment: 05-06-2024
to see stock.
Add to basket
Description
Description
  • Reflection avoided through tilted mount for emission filters
  • Maximum filter thickness: 5mm
Specifications
Specifications
Gooseneck Light Guide, Reflector Module
0.5 cm