Läs mer
Carnitinamide Chloride, TRC
Produktkod. 30363933
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30363933 100 mg
1 options
Produktkod. 30363933 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC521881100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (2R)-4-Amino-2-hydroxy-N,N,N-trimethyl-4-oxobutan-1-aminium Chloride
CAS 6490-20-6
Molekylformel C7 H17 N2 O2 . Cl
Kvantitet 100 mg
Synonym 1-Butanaminium, 4-amino-2-hydroxy-N,N,N-trimethyl-4-oxo-, chloride (1:1), (2R)-, 1-Butanaminium, 4-amino-2-hydroxy-N,N,N-trimethyl-4-oxo-, chloride, (2R)- (9CI), 1-Butanaminium, 4-amino-2-hydroxy-N,N,N-trimethyl-4-oxo-, chloride, (R)-, Ammonium, (3-carbamoyl-2-hydroxypropyl)trimethyl-, chloride, (-)- (8CI), ((3R)-3-Carbamoyl-2-hydroxypropyl)trimethylammonium chloride, Carnitinamide chloride, Carnitine amide chloride, L-(-)-Carnitinamide chloride, (2R)-4-Amino-2-hydroxy-N,N,N-trimethyl-4-oxobutan-1-aminium Chloride, Carnitinamide Chloride, Levocarnitine Imp. C (EP), Levocarnitine EP Impurity C Chloride
InChI formel InChI=1S/C7H16N2O2.ClH/c1-9(2,3)5-6(10)4-7(8)11;/h6,10H,4-5H2,1-3H3,(H-,8,11);1H/t6-;/m1./s1
LEDER [Cl-].C[N+](C)(C)C[C@H](O)CC(=O)N
IUPAC-namn [(2R)-4-amino-2-hydroxy-4-oxobutyl]-trimethylazanium;chloride
Molekylvikt (g/mol) 196.68
Formel vikt 196.0979