Läs mer
Carperidine, LoGiCal
Produktkod. 30461766
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30461766 5 mg
1 options
Produktkod. 30461766 Leverantör LoGiCal Leverantörsnummer LGCFOR1857.01
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Carperidine
CAS 7008-32-4
Analyt- eller komponentnamn Carperidine
Frakt skick Room Temperature
Molekylformel C17 H24 N2 O3
Certifikat och efterlevnad ISO 9001
Synonym 4-Piperidinecarboxylic acid, 1-(3-amino-3-oxopropyl)-4-phenyl-, ethyl ester (9CI), Isonipecotic acid, 1-(2-carbamoylethyl)-4-phenyl-, ethyl ester (8CI), Ethyl 1-(2-carbamoylethyl)-4-phenylpiperidine-4-carboxylate, Carperidine
InChI formel InChI=1S/C17H24N2O3/c1-2-22-16(21)17(14-6-4-3-5-7-14)9-12-19(13-10-17)11-8-15(18)20/h3-7H,2,8-13H2,1H3,(H2,18,20)
Lösningstyp Neat
LEDER CCOC(=O)C1(CCN(CCC(=O)N)CC1)c2ccccc2
IUPAC-namn ethyl 1-(3-amino-3-oxopropyl)-4-phenylpiperidine-4-carboxylate
Industry Type Clinical Testing
Molekylvikt (g/mol) 304.38
Kvantitet 5 mg
Formel vikt 304.1787 g/mol