Läs mer
MedChemExpress Carperitide acetate
Produktkod. 16442058
Encompass_Preferred
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
16442058 5 mg
1 options
Produktkod. 16442058 Leverantör MedChemExpress Leverantörsnummer HYP1235A
MedChemExpress Carperitide acetate (Atrial Natriuretic Peptide (ANP) (1-28), human, porcine acetate) is a 28-amino acid hormone, that is normally produced and secreted by the human heart in response to cardiac injury and mechanical stretch. Carperitide acetate inhibits endothelin-1 secretion in a dose-dependent way.
Specifikationer
Renhet 96.81%
CAS 1366000-58-9
Innehåll och lagring Sealed storage, away from moisture∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
För användning med (applikation) COVID-19-immunoregulation
Molekylvikt (g/mol) 3140.5 g/mol
Produkttyp Peptides
Kvantitet 5 mg
Frakt skick Room temperature
Löslighet H2O : 50 mg/mL (15.92 mM∣Need ultrasonic)
Synonym Atrial Natriuretic Peptide (ANP) (1-28), human, porcine acetate