missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hettich™ Carrier for Hettich Rotor no. 1611
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
Brand: Hettich™ 1643
Additional Details : Weight : 0.10000kg
Beschreibung
- Carrier for Hettich Rotor no. 1611
- For use with ROTOFIX 32 A and Universal 320/320 R Centrifuge, 8-place
Spezifikation
Carrier for Rotor | |
1 x 4 to 10 mL Tube | |
100 mm | |
1 |
Blood/Urine, Round Bottom | |
17 mm | |
Hettich Rotor |