Learn More
Fistreem International™ Cartridge Deionizer
Spare mixed bed resin cartridge suitable for the Cyclon Deionizer. Fistreem International™ Cartridge Deionizer produces a high volume of water between regeneration.
Brand: Fistreem International™ WGS-950-130D
Additional Details : Weight : 1.00000kg
Description
- Useful for providing a suitable supply of deionized water to Cyclon stills.
- Mixed bed resin cartridge produces a high volume of water between regeneration.
Specifications
Cyclon Deionizer/Cyclon Still | |
1 Each | |
Deionizer Cartridge |