Läs mer
Carvacrol-Beta-D-glucuronide, TRC
Produktkod. 30367438
Change view
Kvantitet:
2.5 mg
50 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30367438 2.5 mg 2.5mg
30356548 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30367438 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC1842652.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1642562-02-4
Molekylformel C16H22O7
Molekylvikt (g/mol) 326.34
Synonym 2-Methyl-5-(1-methylethyl)Phenyl-beta-D-glucopyranosiduronic Acid
IUPAC-namn (2S,3S,5R,6S)-3,4,5-trihydroxy-6-(2-methyl-5-propan-2-ylphenoxy)oxane-2-carboxylic acid
LEDER CC(C)c1ccc(C)c(O[C@@H]2O[C@@H]([C@@H](O)C(O)[C@H]2O)C(=O)O)c1
Specifikationer
Kemiskt namn eller material Carvacrol-beta-D-glucuronide
Kvantitet 2.5 mg
InChI formel InChI=1S/C16H22O7/c1-7(2)9-5-4-8(3)10(6-9)22-16-13(19)11(17)12(18)14(23-16)15(20)21/h4-7,11-14,16-19H,1-3H3,(H,20,21)/t11?,12-,13+,14-,16+/m0/s1
Formel vikt 326.1366