Läs mer
Carvedilol Beta-D-Glucuronide (mixture of diasteromers), TRC
Produktkod. 30350349
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30350349 5 mg
1 options
Produktkod. 30350349 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC1846405MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Carvedilol beta-D-Glucuronide (mixture of diasteromers)
CAS 114869-83-9
Molekylformel C30 H34 N2 O10
Kvantitet 5 mg
Synonym Carvedilol-O-Beta-D-Glucuronide
InChI formel InChI=1S/C30H34N2O10/c1-38-21-10-4-5-11-22(21)39-14-13-31-15-17(16-40-30-27(35)25(33)26(34)28(42-30)29(36)37)41-23-12-6-9-20-24(23)18-7-2-3-8-19(18)32-20/h2-12,17,25-28,30-35H,13-16H2,1H3,(H,36,37)/t17?,25-,26-,27+,28-,30+/m0/s1
LEDER COc1ccccc1OCCNCC(CO[C@@H]2O[C@@H]([C@@H](O)[C@H](O)[C@H]2O)C(=O)O)Oc3cccc4[nH]c5ccccc5c34
IUPAC-namn (2S,3S,4S,5R,6R)-6-[2-(9H-carbazol-4-yloxy)-3-[2-(2-methoxyphenoxy)ethylamino]propoxy]-3,4,5-trihydroxyoxane-2-carboxylic acid
Molekylvikt (g/mol) 582.6
Formel vikt 582.2213