Läs mer
Carvedilol Bis-carbazole, TRC
Produktkod. 30356117
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30356117 100 mg
1 options
Produktkod. 30356117 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC184645100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Carvedilol Biscarbazole
CAS 918903-20-5
Molekylformel C39 H39 N3 O6
Kvantitet 100 mg
Synonym 1,1'-[[2-(2-Methoxyphenoxy)ethyl]nitrilo]bis[3-(9H-carbazol-4-yloxy)propan-2-ol], Carvedilol Imp. B (EP), Carvedilol Biscarbazole
InChI formel InChI=1S/C39H39N3O6/c1-45-34-16-6-7-17-35(34)46-21-20-42(22-26(43)24-47-36-18-8-14-32-38(36)28-10-2-4-12-30(28)40-32)23-27(44)25-48-37-19-9-15-33-39(37)29-11-3-5-13-31(29)41-33/h2-19,26-27,40-41,43-44H,20-25H2,1H3
LEDER COc1ccccc1OCCN(CC(O)COc2cccc3[nH]c4ccccc4c23)CC(O)COc5cccc6[nH]c7ccccc7c56
IUPAC-namn 1-(9H-carbazol-4-yloxy)-3-[[3-(9H-carbazol-4-yloxy)-2-hydroxypropyl]-[2-(2-methoxyphenoxy)ethyl]amino]propan-2-ol
Molekylvikt (g/mol) 645.74
Formel vikt 645.2839