Läs mer
Casanthranol (A mixture of Casanthranol A, Casanthranol B, Cascaroside A and Cascaroside B) (Technical Grade), TRC
Produktkod. 30363414
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30363414 500 mg
1 options
Produktkod. 30363414 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC226503500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Casanthranol
CAS 8024-48-4
Molekylformel C21H22O10
Kvantitet 500 mg
Synonym mixture of Casanthranol A, Casanthranol B, Cascaroside A and Cascaroside B
InChI formel InChI=1S/C21H22O10/c22-6-8-4-10-15(12(25)5-8)17(27)14-9(2-1-3-11(14)24)20(10)31-21-19(29)18(28)16(26)13(7-23)30-21/h1-5,13,16,18-26,28-29H,6-7H2/t13-,16+,18-,19-,20-,21+/m1/s1
LEDER C1=CC2=C(C(=C1)O)C(=O)C3=C(C=C(C=C3[C@@H]2O[C@H]4[C@@H]([C@@H]([C@H]([C@H](O4)CO)O)O)O)CO)O
IUPAC-namn (10R)-1,8-dihydroxy-3-(hydroxymethyl)-10-[(2R,3R,4R,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-10H-anthracen-9-one
Molekylvikt (g/mol) 434.4
Formel vikt 434.1213