Beskrivning
CASD1 Polyclonal specifically detects CASD1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifikationer
Specifikationer
| Antigen | CASD1 |
| Användningsområden | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Klassificering | Polyclonal |
| Konjugera | Unconjugated |
| Utspädning | Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Formulering | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Gene Alias | C7orf12O-acetyltransferase, CAS1 domain containing 1, CAS1 domain-containing protein 1, FLJ21213, FLJ21879, FLJ41901, NBLA04196, WUGSC:H_GS542D18.2 |
| Gensymboler | CASD1 |
| Värd art | Rabbit |
| Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:SIAKPHVIVAGAATWSIKIHNGSSEALSQYKMNITSIAPLLEKLAKTSDVYWVLQDPVYEDLLSENRKMITNEKIDAY |
| Visa mer |
For Research Use Only