missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Casein, High Nitrogen, 95%

For research use

Brand:  Alfa Aesar™ J64482.A1

Additional Details : CAS Number : 9000-71-9 Weight : 1.00000kg

 View more versions of this product

Product Code. 15496409

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +46 31 352 32 00 for assistance


Specifications

Specifications

Lyophilized Powder
95%
Bovine milk
Casein, high nitrogen, 95%
Lyophilized powder
1kg
RUO
No
Videos
SDS

For Research Use Only