missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase 2/p18 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10436-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein which is a member of the cysteine-aspartic acid protease (caspase) family. Sequential activation of caspases plays a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes which undergo proteolytic processing at conserved aspartic residues to produce two subunits, large and small, that dimerize to form the active enzyme. The proteolytic cleavage of this protein is induced by a variety of apoptotic stimuli. Alternative splicing of this gene results in multiple transcript variants that encode different isoforms.Specifications
Caspase 2/p18 | |
Polyclonal | |
Unconjugated | |
CASP2 | |
CASP 2, CASP2, Caspase 2, Caspase 2/p18, Caspase2, ICH 1L, ICH 1L/1S, ICH1, NEDD 2, NEDD2, Protease ICH 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12366, 64314, 835 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P29594, P42575, P55215 | |
Casp2 | |
Caspase 2/p18 Fusion Protein Ag0694 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |